4-Methoxy-N-methylbenzylamine 95%

Supplier: Thermo Scientific Chemicals
Warning

45173-0010 45173-0100
TS45173-0010EA 59 USD
TS45173-0010 TS45173-0100
4-Methoxy-N-methylbenzylamine 95%
4-Methoxy-N-methylbenzylamine
Formula: C₉H₁₃NO
Boiling Pt: 222 °C
Density: 1.008 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD04115407
CAS Number: 702-24-9

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5310 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR