tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%

Supplier: Thermo Scientific Chemicals

45172-0050 45172-0250
TS45172-0050EA 389 USD
TS45172-0050 TS45172-0250
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate
Formula: C₁₆H₂₈BNO₄
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD03840345
CAS Number: 286961-14-6

Order Now

Specification Test Results

Appearance White to yellow to tan Powder
Infrared spectrum Conforms
Melting point 114 to 118 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR