3-Bromo-5-fluoropicolinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

45143-0010 45143-0050
TS45143-0010EA 196 USD
TS45143-0010 TS45143-0050
3-Bromo-5-fluoropicolinonitrile 97%
3-Bromo-5-fluoropicolinonitrile
Formula: C₆H₂BrFN₂
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD16988825
CAS Number: 950670-18-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or solid
Infrared spectrum Conforms
Melting point 88 to 92 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR