2-Amino-5-methoxybenzonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45134-0010
45134-0050
TS45134-0010EA
112
USD
TS45134-0010
TS45134-0050
2-Amino-5-methoxybenzonitrile 95%
2-Amino-5-methoxybenzonitrile
Formula:
C₈H₈N₂O MW: 148.1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05149280 CAS Number: 23842-82-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to yellow to brown Powder or low melting solid |
Infrared spectrum | Conforms |
Melting point | 39 to 45 °C |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...