(4-Iodophenyl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Iodophenyl)acetic acid

45090-0010 45090-0050
TS45090-0010EA 62 USD
TS45090-0010 TS45090-0050
(4-Iodophenyl)acetic acid 97%
(4-Iodophenyl)acetic acid
Formula: (C₆H₄I)CH₂COOH
MW: 262.05 g/mol
Melting Pt: 140…141.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00082985
CAS Number: 1798-06-7

Order Now

Specification Test Results

Appearance White to off-white to light brown Powder or solid
Infrared spectrum Conforms
Melting point 137 to 143 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR