4'-Chlorobiphenyl-4-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [1,1'-Biphenyl]-4-carboxaldehyde, 4'-chloro-, 4'-Chlorobiphenyl-4-carbaldehyde, 4'-Chloro-4-biphenylcarbaldehyde, 4'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde

45036-0010 45036-0050
TS45036-0010EA 185 USD
TS45036-0010 TS45036-0050
4'-Chlorobiphenyl-4-carboxaldehyde 97%
4'-Chlorobiphenyl-4-carboxaldehyde
Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD01631911
CAS Number: 80565-30-6

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 112 to 118 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR