[4-(4-Chlorophenyl)phenyl]sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4'-Chloro-[1,1'-biphenyl]-4-sulphonyl chloride, 4'-Chlorobiphenyl-4-sulphonyl chloride

45033-0010 45033-0100
TS45033-0010EA 49 USD
TS45033-0010 TS45033-0100
[4-(4-Chlorophenyl)phenyl]sulfonyl chloride 97%
[4-(4-Chlorophenyl)phenyl]sulfonyl chloride
Formula: C₁₂H₈Cl₂O₂S
MW: 287.17 g/mol
Melting Pt: 109…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631918
CAS Number: 20443-74-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 106 to 112 °C
HPLC ≥96 % as diethylsulfonamide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR