3'-Chloro-4'-hydroxyacetanilide 98%

Supplier: Thermo Scientific Chemicals
Warning

45008-0010 45008-0050
TS45008-0010EA 59 USD
TS45008-0010 TS45008-0050
3'-Chloro-4'-hydroxyacetanilide 98%
3'-Chloro-4'-hydroxyacetanilide
Formula: C₈H₈ClNO₂
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD01318123
CAS Number: 3964-54-3

Order Now

Specification Test Results

Appearance Off-white to light pink to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 146 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR