4-[1,2,4]Triazol-4-ylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4H-1,2,4-Triazol-4-yl)benzoic acid, 4-(p-Carboxyphenyl)-1,2,4-triazole

44996-0010 449960050
TS44996-0010EA 311 USD
TS44996-0010 TS449960050
4-[1,2,4]Triazol-4-ylbenzoic acid 97%
4-[1,2,4]Triazol-4-ylbenzoic acid
Formula: C₉H₇N₃O₂
MW: 189.18 g/mol
Melting Pt: 330 °C
Storage Temperature: Ambient
MDL Number: MFCD06654839
CAS Number: 157069-48-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR