4,5-Difluoro-o-phenylenediamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5-Difluoro-1,2-phenylenediamine, 4,5-Difluorobenzene-1,2-diamine, 1,2-Diamino-4,5-difluorobenzene

44981-0010 44981-0050 44981-0250
TS44981-0010EA 49 USD
TS44981-0010 TS44981-0050 TS44981-0250
4,5-Difluoro-o-phenylenediamine 97%
4,5-Difluoro-o-phenylenediamine
Formula: C₆H₆F₂N₂
Melting Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00061131
CAS Number: 76179-40-3

Order Now

Specification Test Results

Appearance Off-white to red-brown Crystalline powder (May darken on storage)
Infrared spectrum Conforms
GC ≥96.0 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR