1,3-Di-tert-butylbenzene 99%

Supplier: Thermo Scientific Chemicals

44889-0010 44889-0050 44889-0250
TS44889-0010EA 81 USD
TS44889-0010 TS44889-0050 TS44889-0250
1,3-Di-tert-butylbenzene 99%
1,3-Di-tert-butylbenzene
Formula: C₁₄H₂₂
MW: 190.33
Boiling Pt: 106…107 °C (18 mmHg)
Melting Pt: 10…11 °C
Density: 0.86 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008830
CAS Number: 1014-60-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4890 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR