4-Bromo-2,6-dinitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Warning

44886-0010 44886-0050
TS44886-0010EA 85 USD
TS44886-0010 TS44886-0050
4-Bromo-2,6-dinitrotoluene 97%
4-Bromo-2,6-dinitrotoluene
Formula: C₇H₅BrN₂O₄
MW: 261.03 g/mol
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD07357280
CAS Number: 95192-64-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline solid
Infrared spectrum Conforms
Melting point 86 to 89 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR