4-Amino-3-nitrophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amino-3-nitrophenylboronicacid
44530-0010
44530-0050
TS44530-0010EA
315
USD
TS44530-0010
TS44530-0050
4-Amino-3-nitrophenylboronic acid 97%
4-Amino-3-nitrophenylboronic acid
Formula:
C₆H₇BN₂O₄ MW: 181.9418 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07437851 CAS Number: 89466-07-9 |
Specification Test Results
Appearance | Orange to red Solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...