4-Formylbenzeneboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

44504-0050 44504-0250
TS44504-0050EA 92 USD
TS44504-0050 TS44504-0250
4-Formylbenzeneboronic acid pinacol ester 97%
4-Formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₇BO₃
MW: 232.086 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD04972375
CAS Number: 128376-64-7

Order Now

Specification Test Results

Appearance White to off-white to light yellow Solid or crystalline solid
Infrared spectrum Conforms
Melting point 58 to 64 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR