2-Fluoro-5-formylphenylboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-5-formylbenzeneboronic acid pinacol ester, 4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Fluoro-5-formylphenylboronic acid, pinacol ester

44503-0010 44503-0050
TS44503-0010EA 143 USD
TS44503-0010 TS44503-0050
2-Fluoro-5-formylphenylboronic acid pinacol ester 95%
2-Fluoro-5-formylphenylboronic acid pinacol ester
Formula: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
Boiling Pt: 105…113 °C (.1 mmHg)
Melting Pt: 32…37 °C
Storage Temperature: Ambient
MDL Number: MFCD11867877
CAS Number: 1112208-82-8

Order Now

Specification Test Results

Appearance Colorless to yellow Low melting solid or viscous liquid
Infrared spectrum Conforms
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR