4-Amino-2-fluorophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

44449-0010 44449-0050
TS44449-0010EA 163 USD
TS44449-0010 TS44449-0050
4-Amino-2-fluorophenylboronic acid pinacol ester 97%
4-Amino-2-fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BFNO₂
Melting Pt: 163…166 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09951877
CAS Number: 819057-45-9

Order Now

Specification Test Results

Appearance Off-white to brown Solid
Infrared spectrum Conforms
Melting point 161 to 167 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR