4-(4-Chlorophenyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

44446-0010 44446-0050
TS44446-0010EA 140 USD
TS44446-0010 TS44446-0050
4-(4-Chlorophenyl)aniline 97%
4-(4-Chlorophenyl)aniline
Formula: C₁₂H₁₀ClN
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00477989
CAS Number: 135-68-2

Order Now

Specification Test Results

Appearance Off-white to orange to brown Solid
Infrared spectrum Conforms
Melting point 126 to 132 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR