4-Amino-2-chlorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-4-cyanoaniline

44388-0100 44388-0500
TS44388-0100EA 88 USD
TS44388-0100 TS44388-0500
4-Amino-2-chlorobenzonitrile 99%
4-Amino-2-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00035926
CAS Number: 20925-27-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to cream to brown Crystalline powder
Infrared spectrum Conforms
Melting point 116 to 118 °C
Water ≤0.5 %
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR