4-Bromo-o-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

44340-0050 44340-0250
TS44340-0050EA 75 USD
TS44340-0050 TS44340-0250
4-Bromo-o-toluic acid 97%
4-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00040905
CAS Number: 68837-59-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR