5-Methoxyanthranilic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

44334-0010 44334-0050
TS44334-0010EA 64 USD
TS44334-0010 TS44334-0050
5-Methoxyanthranilic acid 97%
5-Methoxyanthranilic acid
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 151…151.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00016509
CAS Number: 6705-03-9

Order Now

Specification Test Results

Appearance Off-white to brown to purple Powder or solid
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR