3-Bromo-5-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride

44266-0050 44266-0250
TS44266-0050EA 75 USD
TS44266-0050 TS44266-0250
3-Bromo-5-(trifluoromethyl)aniline 97%
3-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02
Boiling Pt: 220…223 °C (760 mmHg)
Density: 1.697 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00236205
CAS Number: 54962-75-3

Order Now

Specification Test Results

Appearance Colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR