4,4'-Dimethyldiphenylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p,p'-Ditolylamine, 4,4'-Dimethyldiphenylamine

44259-0050 44259-0250
TS44259-0050EA 85 USD
TS44259-0050 TS44259-0250
4,4'-Dimethyldiphenylamine 97%
4,4'-Dimethyldiphenylamine
Formula: C₁₄H₁₅N
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00059317
CAS Number: 620-93-9

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 77 to 84 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR