1-Benzyloxy-4-bromobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene

44246-0100 44246-0500
TS44246-0100EA 58 USD
TS44246-0100 TS44246-0500
1-Benzyloxy-4-bromobenzene 97%
1-Benzyloxy-4-bromobenzene
Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Boiling Pt: 166…167 °C (4Torr)
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00028016
CAS Number: 6793-92-6

Order Now

Specification Test Results

Appearance White to light brown Powder or needles
Infrared spectrum Conforms
Melting point 59 to 64 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR