4-Bromo-2-methoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

44195-0050 44195-0250
TS44195-0050EA 81 USD
TS44195-0050 TS44195-0250
4-Bromo-2-methoxyphenol 98%
4-Bromo-2-methoxyphenol
Formula: C₇H₇BrO₂
Boiling Pt: 90…92 °C (1hPa)
Melting Pt: 35…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00051937
CAS Number: 7368-78-7

Order Now

Specification Test Results

Appearance Colorless to white to light brown Low melting crystalline solid
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR