3-Fluoro-4-hydroxybenzoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

44189-0010 44189-0050
TS44189-0010EA 77 USD
TS44189-0010 TS44189-0050
3-Fluoro-4-hydroxybenzoic acid 95%
3-Fluoro-4-hydroxybenzoic acid
Formula: C₇H₅FO₃
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00143161
CAS Number: 350-29-8

Order Now

Specification Test Results

Appearance Off-white to brown Powder
Infrared spectrum Conforms
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR