trans-4-Bromo-β-nitrostyrene 99%

Supplier: Thermo Scientific Chemicals
Warning

44016-0010 44016-0050
TS44016-0010EA 44 USD
TS44016-0010 TS44016-0050
trans-4-Bromo-β-nitrostyrene 99%
trans-4-Bromo-β-nitrostyrene
Formula: C₈H₆BrNO₂
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00191856
CAS Number: 5153-71-9

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Melting point 147 to 153 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR