4-Bromophenethyl bromide 96%

Supplier: Thermo Scientific Chemicals
Warning

43953-0010 43953-0100
TS43953-0010EA 44 USD
TS43953-0010 TS43953-0100
4-Bromophenethyl bromide 96%
4-Bromophenethyl bromide
Formula: C₈H₈Br₂
Boiling Pt: 97…98.5 °C (1 mmHg)
Density: 1.73 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00045052
CAS Number: 1746-28-7
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR