(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-1-Benzyl-3-aminopyrrolidine, (S)-(+)-1-Benzyl-3-aminopyrrolidine

43788-0010 43788-0050
TS43788-0010EA 95 USD
TS43788-0010 TS43788-0050
(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
Boiling Pt: 108…115 °C (5 mmHg)
Density: 1.02 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00082766
CAS Number: 114715-38-7
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +1° to +3° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR