1-Benzyloxy-3-bromobenzene 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Benzyloxybromobenzene, 1-(Benzyloxy)-3-bromobenzene

43779-0100 43779-0500
TS43779-0100EA 70 USD
TS43779-0100 TS43779-0500
1-Benzyloxy-3-bromobenzene 96%
1-Benzyloxy-3-bromobenzene
Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00155065
CAS Number: 53087-13-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White Solid
Infrared spectrum Conforms
Melting point 59 to 64 °C
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR