2-Aminopyridine-4-methanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4-pyridinylmethanol, (2-Aminopyridin-4-yl)-methanol, (2-Aminopyridin-4-yl)methanol, (2-Aminopyrid-4-yl)methanol

43737-0010 43737-0050
TS43737-0010EA 82 USD
TS43737-0010 TS43737-0050
2-Aminopyridine-4-methanol 97%
2-Aminopyridine-4-methanol
Formula: C₆H₈N₂O
MW: 124.143 g/mol
Melting Pt: 75.5…84 °C
Storage Temperature: Ambient
MDL Number: MFCD03791261
CAS Number: 105250-17-7

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR