Isoproterenol hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DL-Isoproterenol hydrochloride, (±)-Isoproterenol hydrochloride, Isoprenaline hydrochloride

43721-0050 43721-0250 43721-1000
TS43721-0050EA 57 USD
TS43721-0050 TS43721-0250 TS43721-1000
Isoproterenol hydrochloride 98%
Isoproterenol hydrochloride
Formula: C₁₁H₁₇NO₃
MW: 247.72 g/mol
Melting Pt: 165…175 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012603
CAS Number: 51-30-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 164 to 170 °C
Loss on drying ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR