3-Benzyloxy-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Propanol, 3-(benzyloxy)-, 3-phenylmethoxy-1-propanol, Trimethylene glycol monobenzyl ether, 3-(Benzyloxy)-1-propanol, 3-(Benzyloxy)propan-1-ol

43657-0050 43657-0250
TS43657-0050EA 74 USD
TS43657-0050 TS43657-0250
3-Benzyloxy-1-propanol 97%
3-Benzyloxy-1-propanol
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 111…114 °C (2 mmHg)
Density: 1.049 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00029659
CAS Number: 4799-68-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR