3,5-Dichlorothiophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

43568-0010 43568-0050
TS43568-0010EA 36 USD
TS43568-0010 TS43568-0050
3,5-Dichlorothiophenol 97%
3,5-Dichlorothiophenol
Formula: C₆H₄Cl₂S
Boiling Pt: 125…130 °C (12 mmHg)
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00041423
CAS Number: 17231-94-6
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or solid
Infrared spectrum Conforms
Melting point 62 to 65 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR