4-Bromo-m-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

43478-0050 43478-0250
TS43478-0050EA 67 USD
TS43478-0050 TS43478-0250
4-Bromo-m-toluic acid 97%
4-Bromo-m-toluic acid
Formula: C₈H₇BrO₂
Melting Pt: 215.5…216.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00039526
CAS Number: 7697-28-1

Order Now

Specification Test Results

Appearance Yellow to orange to tan Powder
Infrared spectrum Conforms
Melting point 213 to 217 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR