7-Amino-1-tetralone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Amino-alpha-tetralone, 7-Amino-3,4-dihydro-1(2H)-naphthalenone

43326-0010 43326-0050
TS43326-0010EA 133 USD
TS43326-0010 TS43326-0050
7-Amino-1-tetralone 97%
7-Amino-1-tetralone
Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD07369817
CAS Number: 22009-40-1

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 135 to 140 °C
GC ≥96.0 %
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR