4-Methoxybenzenesulfonamide 95%

Supplier: Thermo Scientific Chemicals
Warning

43291-0050 43291-0250
TS43291-0050EA 74 USD
TS43291-0050 TS43291-0250
4-Methoxybenzenesulfonamide 95%
4-Methoxybenzenesulphonamide
Formula: C₇H₉NO₃S
MW: 187.22 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00025392
CAS Number: 1129-26-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 109 to 113 °C
HPLC ≥94.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.5 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR