6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43271-0010
43271-0050
TS43271-0010EA
209
USD
TS43271-0010
TS43271-0050
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one 97%
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one
Formula:
C₈H₆BrNO₂ Melting Pt: 149…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00694789 CAS Number: 67927-44-0 |
Specification Test Results
Appearance | Off-white Powder |
Infrared spectrum | Conforms |
Melting point | 147 to 151 °C |
GC | ≥96.0 % |
Specific optical rotation | +63° to +67° (20 °C, 589 nm) (c=4, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...