Methyl-3-aminobenzo[b]thiophene-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

43241-0010 43241-0050
TS43241-0010EA 51 USD
TS43241-0010 TS43241-0050
Methyl-3-aminobenzo[b]thiophene-2-carboxylate 97%
Methyl 3-aminobenzo[b]thiophene-2-carboxylate
Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Melting Pt: 109…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00206744
CAS Number: 35212-85-2

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 108 to 112 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR