trans-β-Styrylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: E-Phenylethenylboronic acid, pinacol ester, Trans-2-Phenylvinylboronic acid pinacol ester, trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene

43203-0010 43203-0050
TS43203-0010EA 100 USD
TS43203-0010 TS43203-0050
trans-β-Styrylboronic acid pinacol ester 97%
trans-beta-Styrylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₂
MW: 230.12 g/mol
Boiling Pt: 89…93 °C (.1 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03453666
CAS Number: 83947-56-2

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR