trans-2-(4-Chlorophenyl)vinylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E)-(4-Chlorostyryl)boronic acid

43134-0010 43134-0100
TS43134-0010EA 128 USD
TS43134-0010 TS43134-0100
trans-2-(4-Chlorophenyl)vinylboronic acid 97%
trans-2-(4-Chlorophenyl)vinylboronic acid
Formula: C₈H₈BClO₂
MW: 182.42 g/mol
Melting Pt: 133…134 °C
Storage Temperature: Ambient
MDL Number: MFCD02093767
CAS Number: 154230-29-2

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
pH 5.5 to 6.5 (10% aqueous suspension)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR