2-(4-Fluorophenyl)propan-2-amine 97%

Supplier: Thermo Scientific Chemicals
Danger

43106-0010 43106-0050
TS43106-0010EA 214 USD
TS43106-0010 TS43106-0050
2-(4-Fluorophenyl)propan-2-amine 97%
2-(4-Fluorophenyl)propan-2-amine
Formula: C₉H₁₂FN
Boiling Pt: 40 °C (7mbar)
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07777176
CAS Number: 17797-10-3
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR