3-Chloro-4-methylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

43062-0010 43062-0100
TS43062-0010EA 43 USD
TS43062-0010 TS43062-0100
3-Chloro-4-methylphenylboronic acid 97%
3-Chloro-4-methylphenylboronic acid
Formula: C₇H₈BClO₂
Melting Pt: 228.3…231.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039010
CAS Number: 175883-63-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR