3-Methoxy-5-(trifluoromethyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Methoxy-5-trifluoromethylphenylboronic acid

43043-0010 43043-0050
TS43043-0010EA 180 USD
TS43043-0010 TS43043-0050
3-Methoxy-5-(trifluoromethyl)phenylboronic acid 97%
3-Methoxy-5-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₃
Melting Pt: 158.4…160.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056362
CAS Number: 871332-97-7

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 157 to 162 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR