2-Chloro-5-methoxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-methoxyphenylboronic Acid, 2-Chloro-5-methoxybenzeneboronic acid

43034-0010 43034-0050
TS43034-0010EA 59 USD
TS43034-0010 TS43034-0050
2-Chloro-5-methoxyphenylboronic acid 97%
2-Chloro-5-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD06659858
CAS Number: 89694-46-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 109 to 115 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR