2-Chloro-4-fluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

43019-0010 43019-0050
TS43019-0010EA 83 USD
TS43019-0010 TS43019-0050
2-Chloro-4-fluorophenylboronic acid 97%
2-Chloro-4-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD02684295
CAS Number: 313545-72-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR