7-Methoxy-2-naphthol 97%

Supplier: Thermo Scientific Chemicals
Warning

42887-0050 42887-0250
TS42887-0050EA 71 USD
TS42887-0050 TS42887-0250
7-Methoxy-2-naphthol 97%
7-Methoxy-2-naphthol
Formula: C₁₁H₁₀O₂
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00075494
CAS Number: 5060-82-2

Order Now

Specification Test Results

Appearance White to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 116 to 119 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR