Chloromethyl phenyl sulfone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ((Chloromethyl)sulphonyl)benzene, 1-(Chloromethylsulphonyl)benzene, Phenyl Chloromethyl Sulphone

42874-0050 42874-0250
TS42874-0050EA 108 USD
TS42874-0050 TS42874-0250
Chloromethyl phenyl sulfone 99%
Chloromethyl phenyl sulfone
Formula: C₇H₇ClO₂S
MW: 190.64 g/mol
Melting Pt: 51…53 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007551
CAS Number: 7205-98-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 50 to 54 °C
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure
Free alkali (KOH) ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR