Naphthalene-2-sulfonic acid hydrate 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

42869-0050 42869-0250
TS42869-0050EA 22 USD
TS42869-0050 TS42869-0250
Naphthalene-2-sulfonic acid hydrate 90%, Technical Grade
Naphthalene-2-sulfonic acid hydrate
Formula: C₁₀H₈O₃S·xH₂O
Density: 1.44 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149271
CAS Number: 76530-12-6
UN: 2585
ADR: 8,III

Order Now

Specification Test Results

Appearance Grey Powder
Infrared spectrum Conforms
HPLC ≥88 %
Loss on drying ≤0.5 % (1 g, 105°C, 16 h)
Sulfated ash ≤0.1 % (1 g, 600°C)
Appearance of solution (EP) Passes test
Identification Passes test (EP, USP)
Related substances ≤0.5 % (total impurities)
Specific optical rotation +52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/dilute NH3) (Anhydrous substance)
Acidity (USP) Passes test
Color of solution (USP) Passes test
Chloride (Cl) ≤0.018 %
Sulfate (SO4) ≤0.025 %
Arsenic (As) ≤1 µg/g
Soluble starch, sulfite (USP) Passes test
Dextrin (EP, USP) Passes test
Conductivity ≤20 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR