9-Bromo-9-phenylfluorene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9-Bromo-9-phenyl-9H-fluorene

42861-0010 42861-0050
TS42861-0010EA 51 USD
TS42861-0010 TS42861-0050
9-Bromo-9-phenylfluorene 97%
9-Bromo-9-phenylfluorene
Formula: C₁₉H₁₃Br
MW: 321.23 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00075522
CAS Number: 55135-66-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 99 to 103 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR