Methyl-1-hydroxy-2-naphthoate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Hydroxy-2-naphthoic acid methyl ester, 1-Hydroxy-β-naphthoic acid methyl ester, Methyl 1-hydroxy-2-naphthoate

42847-0010 42847-0050
TS42847-0010EA 48 USD
TS42847-0010 TS42847-0050
Methyl-1-hydroxy-2-naphthoate
Methyl-1-hydroxy-2-naphthoate
Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00043823
CAS Number: 948-03-8

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 74 to 80 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR